Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,1-Dichloroethane
Supelco® Organic Reference Standard, 1,1-Dichloroethane
Catalog # 36967-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1,1-Dichloroethane
Catalog # 36967-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • CAS No.:
    75-34-3
  • Description:
    1,1-Dichloroethane
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H302 - H319 - H335 - H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00013673
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Ethylidene chloride
  • Formula:
    CH₃CHCl₂
  • Molecular weight:
    98,96
  • Flash point:
    –10,0 °C - closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P273 - P301 + P312 - P305 + P351 + P338