

1,1,2-Trichloroethane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-00-5
:
Formula:Cl₂HCCH₂Cl
MDL Number:MFCD00000852
:133.4 g/mol
Merck Index:12,09767
Boiling Pt:110…115 °C (760 mmHg)
Melting Pt:–37 °C
Density:1,43 g/cm³
Storage Temperature:Ambient
EINECS:201-166-9
UN:2810
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4700 to 1.4720 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together