Order Entry
Northern Ireland
ContactUsLinkComponent
1,2,3,4-Butanetetracarboxylic acid for synthesis, Sigma-Aldrich®
Irritant
1,2,3,4-Butanetetracarboxylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
1,2,3,4-Butanetetracarboxylic acid for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    118 g/l (19 °C)
  • Melting Point:
    187 - 190 °C
  • Molar Mass:
    234.16 g/mol

Specification Test Results

Related Information

Frequently Bought Together