Order Entry
Northern Ireland
ContactUsLinkComponent
Citric acid, anhydrous ≥99.5%, crystalline powder for electrophoresis
Irritant
Citric acid, anhydrous ≥99.5%, crystalline powder for electrophoresis
Catalog # ICNA04800681
Supplier:  MP Biomedicals
CAS Number:  
undefined
Irritant
Citric acid, anhydrous ≥99.5%, crystalline powder for electrophoresis
Catalog # ICNA04800681
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Synonyms:
    β-Hydroxytricarballylic Acid; Citreoviridin
  • Form:
    White Crystalline Powder
  • Storage:
    Store at room temperature (15 to 30 °C)
  • Heavy Metals:
    10 ppm
  • pH:
    1 - 3 (5% aq soln) (Lit.)
  • pka:
    (1) 3,13, (2) 4,76, (3) 6,4
  • Residue on Ignition:
    0,05%
  • Water Content:
    0,5%

Specification Test Results

About this item

 :
 :192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1,665 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669