Formic acid 99% ≥99%, BAKER ANALYZED® for LC-MS, tested for UHPLC-MS, J.T.Baker®
: Avantor
: Aminic Acid, Methanoic acid
:64-18-6
:
:HCO₂H
:MFCD00003297
:1.22
:46.03 g/mol
:48
:Ambient
:200-579-1
:1779
:8 II
:GHS05
- Environmentally Preferable:
- Assay:min. 99 %
- Residue after Evaporation:max. 20 ppm
- at 220 nm:max. 0.002
- at 254 nm:max. 0.002
- Positive ESI-MS Sensitive Impurities (as Reserpine):max. 50 ppb
- Aluminium (Al):max. 50
- Calcium (Ca):max. 50
- Iron (Fe):max. 50
- Potassium (K):max. 50
- Magnesium (Mg):max. 50
- Sodium (Na):max. 50
- at 240 nm:max. 0.1
- at 260 nm:max. 0.01
Frequently Bought Together