
Margaric acid 98%
:506-12-7
:
:C₁₇H₃₄O₂
:MFCD00002751
:270.46 g/mol
:13,05775
:364 °C (760 mmHg)
:59…61 °C
:1,02 g/cm³ (20 °C)
:208-027-1
:GHS07
:Ambient
- Appearance:White to off-white Glistening crystalline powder or flakes
- Infrared spectrum:Conforms
- Melting point:59 to 63 °C
- GC:≥97.5 %
Frequently Bought Together