Order Entry
Northern Ireland
ContactUsLinkComponent
Propyl acetate for synthesis, Sigma-Aldrich®
Flammable
Irritant
Propyl acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Propyl acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    21.2 g/l (20 °C)
  • Melting Point:
    -95 °C
  • Molar Mass:
    102.13 g/mol
  • Boiling Point:
    101.5 °C (1013 hPa)
  • Vapor Pressure:
    33 hPa (20 °C)
  • Flash Point:
    14 °C
  • Explosion Limit:
    1.7 - 8 %(V)
  • Density:
    0.89 g/cm3 (20 °C)
  • Ignition Point:
    430 °C

Specification Test Results

Related Information

Frequently Bought Together