Order Entry
Northern Ireland
ContactUsLinkComponent
Amyl acetate ≥98.0% (by GC), EMPLURA®, Supelco®
Flammable
Amyl acetate ≥98.0% (by GC), EMPLURA®, Supelco®
Catalog # 8.18700.1000
Supplier:  Merck
CAS Number:  
undefined
Flammable
Amyl acetate ≥98.0% (by GC), EMPLURA®, Supelco®
Catalog # 8.18700.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (GC, area%):
    ≥ 98.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    0.875 - 0.877
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :130.19 g/mol
Boiling Pt:149 °C (1013 hPa)
Melting Pt:–71 °C
Density:0,876 g/cm³ (20 °C)
Flash Pt:23 °C
Storage Temperature:Ambient
EINECS:211-047-3
UN:1104
ADR:3,III
 :M-Clarity™ quality level = MQ200
 :GHS02
CAS Number:628-63-7
Formula:C₇H₁₄O₂
MDL Number:MFCD00009500