Order Entry
Northern Ireland
ContactUsLinkComponent
Methyl acetate
Methyl acetate
Catalog # 8.19244.6190
Supplier:  Merck
CAS Number:  
Methyl acetate
Catalog # 8.19244.6190
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    190 L
  • Seq:
    5514

Specifications

About this item

 :
Merck Index:13,06038
Boiling Pt:56,9 °C (1013 hPa)
Melting Pt:–98 °C
Density:0,938 g/cm³ (20 °C)
Flash Pt:–13 °C
Storage Temperature:Ambient
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :74.08 g/mol
EINECS:201-185-2
UN:1231
 :GHS02
 :GHS07
CAS Number:79-20-9
Formula:C₃H₆O₂
MDL Number:MFCD00008711