Order Entry
Northern Ireland
ContactUsLinkComponent
2-Hexanone, Supelco®
Flammable
Health
Irritant
2-Hexanone, Supelco®
Catalog # 02473-5ML
Supplier:  Merck
CAS Number:  
undefined
Flammable
Health
Irritant
2-Hexanone, Supelco®
Catalog # 02473-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :100.16 g/mol
Merck Index:13,06060
Boiling Pt:127 °C (1013 hPa)
Melting Pt:–56,9 °C
Density:0,812 g/cm³ (20 °C)
Flash Pt:31 °C
Storage Temperature:Ambient
EINECS:209-731-1
UN:1224
ADR:3,III
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
 :GHS07
CAS Number:591-78-6
Formula:C₆H₁₂O
MDL Number:MFCD00009482