


2-Hexanone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:591-78-6
:
Formula:C₆H₁₂O
MDL Number:MFCD00009482
:100.16 g/mol
Merck Index:14,6033
Boiling Pt:126…127 °C
Melting Pt:–56 °C
Density:0,811
Flash Pt:23 °C (73 °F)
Storage Temperature:Ambient
EINECS:209-731-1
UN:1224
ADR:3,III
:GHS08
:GHS02
:GHS07
Related Information
Frequently Bought Together