Triethylammonium dihydrogentrifluoride 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride
CAS Number:73602-61-6
:
Formula:C₆H₁₈F₃N
MDL Number:MFCD00043294
Boiling Pt:70 °C (15 mmHg)
Melting Pt:–29 °C
Density:0.989 g/cm³
Molecular Weight:161.21 g/mol
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:277-550-5
UN:2922
ADR:8 I
:GHS06
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Titration with NaOH:20 to 25 % (available HF)
Specification Test Results
Frequently Bought Together