trans-2-Aminocyclohexanol hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5456-63-3
:
Formula:C₆H₁₄ClNO
MDL Number:MFCD00001490
Melting Pt:172…175 °C
:151.64 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together