

Ethyl acrylate stabilised, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:140-88-5
:
Formula:C₅H₈O₂
MDL Number:MFCD00009188
:100.12 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ200
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:205-438-8
UN:1917
:GHS02
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:15 g/l (20 °C)
- Melting Point:-71 °C
- Molar Mass:100.11 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:39 hPa (20 °C)
- Flash Point:8 °C
- Refractive Index:1.4049 (20 °C, 589 nm)
- Explosion Limit:1.8 - 14 %(V)
- Density:0.92 g/cm3 (20 °C)
- Ignition Point:355 °C DIN 51794
- Water absorption:124 g/kg (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together