

Osmium(III) chloride hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14996-60-2
:
Formula:Cl₃Os·H₂O
MDL Number:MFCD00149815
:314.6 g/mol
Melting Pt:>500 °C (dec.)
Storage Temperature:Ambient
EINECS:236-587-7
UN:2923
ADR:8,III
:GHS06
:GHS05
Specification Test Results
- Appearance:Dark red to black Powder or crystals
- Osmium (Os):53 to 56 %
Specification Test Results
Frequently Bought Together