

Osmium(III) chloride trihydrate ≥99.99% (metals basis) Os 52-56%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:135296-80-9
:
Formula:Cl₃Os·H₂O
MDL Number:MFCD00011148
:314.6 g/mol
Melting Pt: Dec.
Storage Temperature:Ambient
EINECS:236-587-7
UN:3260
ADR:8,III
:GHS05
:GHS07
Related Information
Frequently Bought Together