Order Entry
Northern Ireland
ContactUsLinkComponent
N,N-Dimethylformamide
Flammable
Health
Irritant
N,N-Dimethylformamide
Catalog # HYPETS-20672
CAS Number:  
undefined
Flammable
Health
Irritant
N,N-Dimethylformamide
Catalog # HYPETS-20672
CAS Number:  

Specifications

  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:57,5 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284