Order Entry
Northern Ireland
ContactUsLinkComponent
2-(2-Butoxyethoxy)ethyl acetate, Sigma-Aldrich®
2-(2-Butoxyethoxy)ethyl acetate, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-(2-Butoxyethoxy)ethyl acetate, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    64 g/l (20 °C)
  • Melting Point:
    -32 °C
  • Molar Mass:
    204.26 g/mol
  • Boiling Point:
    244 - 250 °C (1013 hPa)
  • Vapor Pressure:
     - 0.01 hPa (20 °C)
  • Flash Point:
    102 °C
  • Refractive Index:
    1.4262 (20 °C, 589 nm)
  • Explosion Limit:
    1.0 - 5.3 %(V)
  • Density:
    0.98 g/cm3 (20 °C)
  • pH:
    3 - 4 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    290 °C

Specification Test Results

Related Information

Frequently Bought Together