Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®
Catalog # 8.14528.0100
Supplier:  Merck
CAS Number:  
Irritant
Bis(8-hydroxyquinolinium)sulphate monohydrate, Sigma-Aldrich®
Catalog # 8.14528.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    177 °C
  • Molar Mass:
    406.41 g/mol
  • pH:
    3.2 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :388.4 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:205-137-1
 :GHS07
CAS Number:207386-91-2
Formula:C₁₈H₁₆N₂O₆S·1H₂O
MDL Number:MFCD00149610