Order Entry
Northern Ireland
ContactUsLinkComponent
Cytochalasin D (from Zygosporium mansoni), Calbiochem®
Cytochalasin D (from Zygosporium mansoni), Calbiochem®
Catalog # 250255-1
CAS Number:  
Cytochalasin D (from Zygosporium mansoni), Calbiochem®
Catalog # 250255-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :507.63 g/mol
EINECS:244-804-1
UN:1544
ADR:6.1,II
CAS Number:22144-77-0
Formula:C₃₀H₃₇NO₆
MDL Number:MFCD00077706