
Ammonium hexachloroosmate(IV) (≥42,5% Os) ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12125-08-5
:
Formula:H₈Cl₆N₂Os
MDL Number:MFCD00010883
Merck Index:14,536
Melting Pt:170 °C (subl.)
Density:2,9
:439.02 g/mol
EINECS:235-188-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together