(±)-Camphor-10-sulphonic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5872-08-2
:
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00074827
:232.3
Melting Pt:>195 °C
Storage Temperature:Ambient
EINECS:227-527-0
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light beige Powder or granular powder
- Infrared spectrum:Conforms
- Loss on drying:≤1.5 % (105°C)
- Titration with NaOH:97.5 to 102.5 % (On dried substance)
- Clarity of solution:(5% in water) Clear solution
- Residue after ignition:≤0.2 %
- Specific optical rotation:-15° to -22° (20°C, 589 nm) (1% in water)
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤500 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤20 ppm
Specification Test Results
Frequently Bought Together