
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride ≥97%
Catalog # H26637.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:176225-09-5
:
Formula:C₇H₃ClF₄O₂S
:262.61 g/mol
Melting Pt:48…51 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
MDL Number:MFCD01091000