4-(Hydroxymethyl)benzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3006-96-0
:
Formula:C₈H₈O₃
MDL Number:MFCD00017598
:152.15 g/mol
Melting Pt:179…183 °C
:GHS07
Related Information
Frequently Bought Together