
3-Bromo-4-hydroxybenzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2973-78-6
:
Formula:C₇H₅BrO₂
MDL Number:MFCD00017348
:201.02 g/mol
Melting Pt:125…128 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together