
3,4-Dibutoxy-3-cyclobutene-1,2-dione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2892-62-8
:
Formula:C₁₂H₁₈O₄
MDL Number:MFCD00037150
:226.27 g/mol
Boiling Pt:155…158 °C (3 mmHg)
Density:1,047
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together