
2-Methoxy-4-nitrobenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21320-91-2
:
Formula:C₇H₆ClNO₅S
MDL Number:MFCD03094697
:251.65 g/mol
Melting Pt:96…99 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Related Information
Frequently Bought Together