

2-Chloro-4-methyl-3-nitropyridine ≥97%
Catalog # H26057.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:23056-39-5
:
Formula:C₆H₅ClN₂O₂
MDL Number:MFCD00012347
Melting Pt:52…54 °C
Flash Pt:>110 °C (230 °F)
UN:2811
ADR:6.1,III
:172.57 g/mol
:GHS06
:GHS07
Storage Temperature:Ambient