
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine ≥97%
Catalog # H32055.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:260.44 g/mol
Boiling Pt:80 °C (32 mmHg)
:GHS07
Storage Temperature:Ambient
CAS Number:75806-84-7
Formula:C₆H₂BrClF₃N
MDL Number:MFCD00153072