Order Entry
Northern Ireland
ContactUsLinkComponent
2-Aminobenzenesulphonamide
2-Aminobenzenesulphonamide
Catalog # MOLE15813553-25G
Supplier:  Molekula
CAS Number:  
2-Aminobenzenesulphonamide
Catalog # MOLE15813553-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :172.21 g/mol
EINECS:221-988-1
CAS Number:3306-62-5
Formula:C₆H₈N₂O₂S
MDL Number:MFCD00007932