4-(Methylamino)phenol sulphate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2794.0500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 101.5 %
- Residue after Ignition:max. 0.1 %
- Suitability for Phopshate Determination:passes test
Specification Test Results
About this item
Synonyms: p-(Methylamino)phenol sulphate, Metol, 4-Methylaminophenol sulphate, 4-(Methylamino)phenol hemisulphate, 4-Methylaminophenolsulphate, 4-(Methylamino)phenol hemisulphate salt
:
:344.39 g/mol
Merck Index:13,06046
Boiling Pt:227 °C (1013 hPa)
Melting Pt:86 °C
Density:1,434 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-237-1
UN:2811
ADR:6.1,III
:GHS08
:GHS07
:GHS09
CAS Number:55-55-0
:51-72-9
Formula:C₁₄H₂₀N₂O₆S
MDL Number:MFCD00013140