Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
4-Cyanobenzophenone ≥98%

4-Cyanobenzophenone ≥98%

4-Cyanobenzophenone ≥98%
Certificates
4-Cyanobenzophenone ≥98%
4-Cyanobenzophenone ≥98%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 98% CAS No.: 1503-49-7 Molecular Formula: C14H9NO Formula Weight: 207.23 Melting Point: 110-114° Beilstein Reference: 641181 UN No.: 3439 MDL No.: MFCD00016382

Synonyms:  4-Benzoylbenzonitrile
  • CAS Number:1503-49-7
  • Formula:C₁₄H₉NO
  • MDL Number:MFCD00016382
  • Molecular Weight:207.23 g/mol
  • Boiling Pt:363 °C (1013 hPa)
  • Melting Pt:110…114 °C
  • Storage Temperature:Ambient
  • EINECS:216-126-6
  • UN:3439
  • ADR:6.1,III
Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.