
2,3-Difluoro-4-hydroxybenzonitrile 95%
Catalog # H32231.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:126162-38-7
:
Formula:C₇H₃F₂NO
MDL Number:MFCD00269596
:155.1 g/mol
Melting Pt:145…149 °C
:GHS07
Storage Temperature:Ambient