Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Chloro-3'-(trifluoromethyl)acetophenone
4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%
4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%
4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%
L19884.06
:
Thermo Fisher Scientific
:
4'-Chloro-3'-(trifluoromethyl)acetophenone ≥99%
L19884.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
222.59 g/mol
:
60…63 °C
:
GHS07
:
Ambient
:
129825-11-2
:
C₉H₆ClF₃O
:
MFCD03094146