Order Entry
Northern Ireland
ContactUsLinkComponent
4-Bromo-2-(trifluoromethoxy)phenol ≥98%
4-Bromo-2-(trifluoromethoxy)phenol ≥98%
Catalog # H31726.06
CAS Number:  
4-Bromo-2-(trifluoromethoxy)phenol ≥98%
Catalog # H31726.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:690264-39-2
 :
Formula:C₇H₄BrF₃O₂
 :257.01 g/mol
Melting Pt:51…55 °C
 :GHS06
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD09037798