Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chloro-5-fluoroaniline
2-Chloro-5-fluoroaniline
Catalog # MOLE74229030-5G
Supplier:  Molekula
CAS Number:  
2-Chloro-5-fluoroaniline
Catalog # MOLE74229030-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :145.56 g/mol
UN:2811
ADR:6.1,III
CAS Number:452-83-5
Formula:C₆H₅ClFN
MDL Number:MFCD00084852