L(-)-Proline for SILAC
Catalog # PIER88211
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:115 mg
- Seq:2465
Specifications
About this item
:
:115.13 g/mol
Merck Index:13,07871
Boiling Pt:245 °C (1013 hPa)
Melting Pt:220…222 °C
Density:1,35 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:205-702-2
CAS Number:147-85-3
Formula:C₅H₉NO₂
MDL Number:MFCD00064318