Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®
 :  Merck
 
 
(1S)-(+)-Camphor-10-sulphonic acid, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    195 °C decomposes
  • Molar Mass:
    232.30 g/mol
  • pH:
    0.3 (200 g/l, H2O)

 

 

Frequently Bought Together