(1R)-(-)-Camphor-10-sulphonic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35963-20-3
:MFCD00064158
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00150753
Boiling Pt:375 °C (1013 hPa)
Melting Pt:∼ 200 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:252-817-9
UN:3261
ADR:8,III
:232.3 g/mol
:GHS05
Related Information
Frequently Bought Together