(1R)-(-)-Camphor-10-sulphonic acid
Catalog # MOLE15982169-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Boiling Pt:375 °C (1013 hPa)
Melting Pt:193…196 °C
Density:1 g/cm³ (20 °C)
:232.3 g/mol
EINECS:252-817-9
CAS Number:35963-20-3
:MFCD00064158
Formula:C₁₀H₁₆O₄S
MDL Number:MFCD00150753