
Dibromomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-95-3
:
Formula:Br₂CH₂
MDL Number:MFCD00000168
:173.83 g/mol
Merck Index:14,6061
Boiling Pt:96…98 °C
Melting Pt:–53 °C
Density:2,495
UN:2664
ADR:6.1,III
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:200-824-2
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together