1-Bromopentane 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:110-53-2
- Formula:BrH₂C(CH₂)₃CH₃
- MDL Number:MFCD00000267
- Molecular Weight:151.05 g/mol
- Merck Index:13,00606
- Boiling Pt:129 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1,21 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-776-0
- UN:1993
- ADR:3,III
2 Options Available Below
Product Details & Documents
- CAS Number:110-53-2
- Formula:BrH₂C(CH₂)₃CH₃
- MDL Number:MFCD00000267
- Molecular Weight:151.05 g/mol
- Merck Index:13,00606
- Boiling Pt:129 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:1,21 g/cm³
- Flash Pt:79 °C
- Storage Temperature:Ambient
- EINECS:203-776-0
- UN:1993
- ADR:3,III
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4430 to 1.4460 (20°C, 589 nm)
- GC:≥97.5 %



