1,9-Dibromononane
Catalog # 8.41727.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 ml
- Seq:2275
Specifications
About this item
:
:286.05 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:–10 °C
Density:1,41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:224-913-0
:GHS07
CAS Number:4549-33-1
Formula:BrH₂C(CH₂)₇CH₂Br
MDL Number:MFCD00000279