1,8-Dibromooctane 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4549-32-0
:
Formula:BrH₂C(CH₂)₆CH₂Br
MDL Number:MFCD00000277
:272.02 g/mol
Boiling Pt:270…272 °C (760 mmHg)
Melting Pt:15…16 °C
Density:1,46 g/cm³
Flash Pt:>112 °C
Storage Temperature:Ambient
EINECS:224-912-5
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4981 to 1.5001 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together