


1,3-Dibromopropane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-64-8
:
Formula:BrH₂CCH₂CH₂Br
MDL Number:MFCD00000255
:201.89 g/mol
Merck Index:12,09844
Boiling Pt:167 °C (1013 hPa)
Melting Pt:–34 °C
Density:1,99 g/cm³ (20 °C)
EINECS:203-690-3
UN:1993
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:47 °C
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:1.68 g/l (30 °C)
- Melting Point:-34 °C
- Molar Mass:201.88 g/mol
- Boiling Point:166 - 168 °C (1013 hPa)
- Vapor Pressure:2.6 hPa (20 °C)
- Flash Point:54 °C
- Refractive Index:1.5233 (20 °C, 589 nm)
- Density:1.98 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together