Order Entry
Northern Ireland
ContactUsLinkComponent
Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Oxidizing
Irritant
Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Catalog # 15290.18
CAS Number:  
Oxidizing
Irritant
Europium trinitrate hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Catalog # 15290.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :446.07 g/mol
Merck Index:14,3904
Melting Pt:85 °C
Density:2,258 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-380-3
UN:1477
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:10031-53-5
Formula:EuN₃O₉·6H₂O
MDL Number:MFCD00149700