


Cerium trinitrate hexahydrate 99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-41-4
:
Formula:Ce(NO₃)₃·6H₂O
MDL Number:MFCD00149631
:434.22 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:39 °C
Density:2,377 g/cm³ (20 °C)
EINECS:233-297-2
UN:1477
ADR:5.1,III
:GHS05
:GHS03
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Powder or crystals and/or chunks
- Assay:≥99.4 % (CeO2/TREO basis)
- Sulfate (SO4):≤200 ppm
- Calcium oxide (CaO):≤50 ppm
- Iron(III)oxide (Fe2O3):≤35 ppm
Specification Test Results
Related Information
Frequently Bought Together