


Lanthanum(III) chloride heptahydrate 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-84-0
:
Formula:LaCl₃·7H₂O
MDL Number:MFCD00149756
Boiling Pt:860 °C (1013 hPa)
Melting Pt:91 °C (dec.)
Storage Temperature:Ambient
EINECS:233-237-5
UN:1759
ADR:8,III
:371.37 g/mol
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:White Crystalline powder or crystals and/or chunks
- Total conc. of imp.as REO:≤0.01 %
Specification Test Results
Frequently Bought Together