


Thallium(III) nitrate trihydrate ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13453-38-8
:
Formula:N₃O₉Tl·3H₂O
MDL Number:MFCD00149874
:444.44 g/mol
Melting Pt:102…105 °C
Density:3,31…3.37 g/cm³ (20 °C)
EINECS:237-325-4
UN:2727
ADR:6.1,II
:GHS06
:GHS03
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together