Aluminium nitrate nonahydrate 98.0-102.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7784-27-2
:
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00149132
:375.13 g/mol
Merck Index:14,352
Melting Pt:73,5 °C
Density:1,72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-751-8
UN:1438
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together